Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 1270 mm, to connect flow sensor to injection valve
Katalog # HEWLG1375-87310
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1375-87310
Capillaries for LC systems
Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 1270 mm, to connect flow sensor to injection valve
Price
1.150,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 1270 mm, to connect flow sensor to injection valve
  • Seq:
    111