Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
  HEWLG1375-87306
Capillaries for LC systems
  HEWLG1375-87306
Capillaries for LC systems
Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
1.150,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
  • Seq:
    108