Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
HEWLG1375-87306
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries, PEEK coated fused silica, B/C, 550 mm, I.Ø 2540 mm, to connect flow sensor to injection valve
- Seq:108