Capillaries for LC systems
Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 2540 mm, to connect EMPV to flow sensor
Katalog # HEWLG1375-87305
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, PEEK coated fused silica, B/B, 220 mm, I.Ø 2540 mm, to connect EMPV to flow sensor
- Seq:107
- Cat. No.:HEWLG1375-87305