Capillaries for LC systems
Capillaries, male to female, stainless steel, 0,6 mm, I.Ø 367 mm, to connect EMPV1 to union
Katalog # HEWLG1361-67306
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, male to female, stainless steel, 0,6 mm, I.Ø 367 mm, to connect EMPV1 to union
- Seq:21