Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, male to female, stainless steel, 40 mm, I.Ø 0,6 mm, to connect union to mixer
Katalog # HEWLG1361-67305
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1361-67305
Capillaries for LC systems
Capillaries for LC systems
Capillaries, male to female, stainless steel, 40 mm, I.Ø 0,6 mm, to connect union to mixer
Price
843,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, male to female, stainless steel, 40 mm, I.Ø 0,6 mm, to connect union to mixer
  • Seq:
    23