Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
Katalog # HEWLG1361-67304
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1361-67304
Capillaries for LC systems
Capillaries for LC systems
Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
Price
843,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
  • Seq:
    22