Capillaries for LC systems
Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
Katalog # HEWLG1361-67304
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, male to female, stainless steel, 111 mm, I.Ø 0,6 mm, to connect union to EMPV2
- Seq:22