Capillaries for LC systems
Capillaries, male to male, stainless steel, 400 mm, I.Ø 0,6 mm, to connect EMPV to next module
HEWLG1361-67302
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries, male to male, stainless steel, 400 mm, I.Ø 0,6 mm, to connect EMPV to next module
- Seq:27