Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, male to male, stainless steel, 175 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
Catalog # HEWLG1361-67301
Capillaries for LC systems
Catalog # HEWLG1361-67301
Capillaries for LC systems
Capillaries for LC systems
Capillaries, male to male, stainless steel, 175 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
Price
843,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Beskrivelse:
    Capillaries, male to male, stainless steel, 175 mm, I.Ø 0,6 mm, to connect pump to 1 multi assembly
  • Seq:
    26