Orders LinkContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Frit adapter, PTFE, 4 mm
Catalog # HEWLG1361-23204
CrossLab Bottle Head Assemblies for HPLC
Catalog # HEWLG1361-23204
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Frit adapter, PTFE, 4 mm
Price
150,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifications
  • Beskrivelse:
    Frit adapter, PTFE, 4 mm
  • Seq:
    13