Capillaries for LC systems
Capillaries, C, stainless steel, to connect needle seat to injection valve
Katalog # HEWLG1329-87101
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, C, stainless steel, to connect needle seat to injection valve
- Seq:20