Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Preparative flow cell, 0,06 mm, 20 bar, quartz
Katalog # HEWLG1315-60018
Leverandør:   AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60018
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Preparative flow cell, 0,06 mm, 20 bar, quartz

Price
30.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Preparative flow cell, 0,06 mm, 20 bar, quartz
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.