Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Preparative flow cell, 0,3 mm, 20 bar, quartz
Katalog # HEWLG1315-60017
Leverandør:   AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60017
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Preparative flow cell, 0,3 mm, 20 bar, quartz

Price
30.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Preparative flow cell, 0,3 mm, 20 bar, quartz
  • Seq:
    05