Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Preparative flow cell, 3 mm, 120 bar, stainless steel for G1315A/B, G1365A/B
Katalog # HEWLG1315-60016
Leverandør: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60016
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Preparative flow cell, 3 mm, 120 bar, stainless steel for G1315A/B, G1365A/B