Capillary column ferrules for gas chromatography
SilTite metal ferrules, for use with 1/16" OD stainless steel tubing includes 2 column nuts
Catalog # HEWL5184-3571
Supplier: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Catalog # HEWL5184-3571
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:SilTite metal ferrules, for use with 1/16" OD stainless steel tubing includes 2 column nuts
- Seq:22