Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,05, 0,25 mm
Catalog # HEWL500-2114
Supplier: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Catalog # HEWL500-2114
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:General purpose graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,05, 0,25 mm
- Seq:01