Orders LinkContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Glass filter, solvent inlet, 40 µm
Katalog # HEWL3150-0944
Leverandør:   AGILENT HEWLETT PACKARD
CrossLab Bottle Head Assemblies for HPLC
Katalog # HEWL3150-0944
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Glass filter, solvent inlet, 40 µm
Price
560,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifikationer
  • Beskrivelse:
    Glass filter, solvent inlet, 40 µm
  • Seq:
    15