pH-elektroder
pH-kombinationselektrode SE 103, glashus, længde 165 mm, SE 103 N, 0...14, −5...+100 °C, Glas, 3 mol/l KCl, Ceramic, 12×170 mm, DIN, Laboratory purposes, industrial production, processing systems
Catalog # 662-3531
Supplier: KNICK
Jump to:
Product Details & Documents
Specifications
- Tilslutning:DIN
- Ø×L:12×170 mm
- Til:Laboratory purposes, industrial production, processing systems
- Model:SE 103 N
- Beskrivelse:pH-kombinationselektrode SE 103, glashus, længde 165 mm
- Materiale:Glas
- Seq:05
- Membran:Ceramic
- Elektrolyt:3 mol/l KCl
- Temperatur:−5...+100 °C
- pH:0...14