Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides with PTFE coating, hvid, 12, 5 mm
Catalog # 631-9423
Supplier:   Epredia
Cavity slides
Catalog # 631-9423
Supplier:  Epredia
Cavity slides
Cavity slides
Cavity slides with PTFE coating, hvid, 12, 5 mm
Price
1.550,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
  • Adhesive surface and hydrophobic mask
  • Hydrophobic epoxy- or PTFE-coating
  • (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).
Specifications
  • Ant. brønde:
    12
  • Farve:
    hvid
  • Beskrivelse:
    Cavity slides with PTFE coating
  • Seq:
    022
  • Brønd Ø:
    5 mm