Slide folders
Slide folder with lock, beige/sort, Pap, 20
Catalog # 631-9006
Supplier: BERGMANN KARTON
Jump to:
Product Details & Documents
These slide folders with a table of content are designed for 20 microscopy slides.
Specifications
- Antal objektglas:20
- Farve:beige/sort
- Beskrivelse:Slide folder with lock
- Materiale:Pap