Dækglas, Menzel-Gläser
Cover glasses, 22×50 mm, #1
Katalog # 631-1365
Leverandør: Epredia
Gå til:
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifikationer
- Antal:#1
- Beskrivelse:Cover glasses
- L×B:22×50 mm
- Seq:12