Linen testers
8×, 17,6 mm, 20×20 mm
Katalog # 630-8322
Leverandør: ESCHENBACH OPTIK
Gå til:
Product Details & Documents
Satin matt chrome plated brass frame with silicate glass lens.
Specifikationer
- Ø:17,6 mm
- Forstørrelse:8×
- B×D:20×20 mm