Indslagslupper
Foldelup, aplanatisk, 8×, 23 mm
Catalog # 630-8232
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
Specifications
- Ø:23 mm
- Forstørrelse:8×
- Beskrivelse:Foldelup, aplanatisk
- Seq:001