Indslagslupper
Foldelup, akromatisk, 10×, 17 mm
Catalog # 630-8221
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
Specifications
- Ø:17 mm
- Forstørrelse:10×
- Beskrivelse:Foldelup, akromatisk
- Seq:002