Orders LinkContactUsLinkComponent
Binocular magnifiers
Binocular magnifiers
Binocular magnifying lens with 180 mm working distance, 2,5×
Katalog # 630-8215
Leverandør:   ESCHENBACH OPTIK
Binocular magnifiers
Katalog # 630-8215
Leverandør:  ESCHENBACH OPTIK
Binocular magnifiers
Binocular magnifiers
Binocular magnifying lens with 180 mm working distance, 2,5×
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
Leveringsinformation: Supplied with a protective box.
Advarsel: Not suitable for spectacle wearers.
Specifikationer
  • Forstørrelse:
    2,5×
  • Beskrivelse:
    Binocular magnifying lens with 180 mm working distance
  • Seq:
    02