Reading magnifiers
Magnifier, hand-held, 2×, 100 mm
Katalog # 630-8151
Leverandør: ESCHENBACH OPTIK
Gå til:
Product Details & Documents
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
- Additional plano-convex lens in the handle for 5× magnification
- Eyelet in the handle, round-the-neck carry cord supplied
- Antistatic coating
Specifikationer
- Ø:100 mm
- Forstørrelse:2×
- Beskrivelse:Magnifier, hand-held
- Seq:01