Penetration probes, K-Type
Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~5 s, −65…+550 °C
Response time (T90): water 0,4 m/s ~5 s, −65…+550 °C
Katalog # 620-4708
Leverandør: Greisinger
Gå til:
Product Details & Documents
Specifikationer
- Område:−65…+550 °C
- Beskrivelse:Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~5 s - Seq:03