Orders LinkContactUsLinkComponent
Standard microliter syringes
Standard microliter syringes
500 µl, 1
Katalog # 613-1720
Leverandør:   Hospidex
Standard microliter syringes
Katalog # 613-1720
Leverandør:  Hospidex
Standard microliter syringes
Standard microliter syringes
500 µl, 1
Price
643,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Providing ultra-precise analyses. The plunger extends down into the tip of the needle ensuring that no dead volume is created.
The bevelled needle tip is 50 mm long.
Specifikationer
  • Spidstype:
    1
  • Volumen:
    500 µl