Transfer pipettes, fine tip
Extended fine tip, mini, 1,0 ml, 20 µl, 104 mm, +, Bulk
Katalog # 612-1787
Leverandør: Alpha Laboratories
Gå til:
Product Details & Documents
Non toxic, food grade LDPE.
- For careful control and small volume transfers
- Ideal for supernatant removal, slide preparation and gel loading
Documents
Specifikationer
- Pakket:Bulk
- Steril:+
- Kapacitet:1,0 ml
- Beskrivelse:Extended fine tip, mini
- Længde:104 mm
- Seq:04
- Dråbevol.:20 µl