Transfer pipettes, fine tip
Fine tip, 3,3 ml, 18 µl, 147 mm, -, Bulk
Katalog # 612-1759
Leverandør: Alpha Laboratories
Gå til:
Product Details & Documents
Non toxic, food grade LDPE.
- For careful control and small volume transfers
- Ideal for supernatant removal, slide preparation and gel loading
Documents
Specifikationer
- Pakket:Bulk
- Steril:-
- Kapacitet:3,3 ml
- Beskrivelse:Fine tip
- Længde:147 mm
- Seq:09
- Dråbevol.:18 µl