Transfer pipettes, fine tip
Extended fine tip, 3,3 ml, 20 µl, 153, -, Bulk
612-1751
Non toxic, food grade LDPE.
- For careful control and small volume transfers
- Ideal for supernatant removal, slide preparation and gel loading
- Pakket:Bulk
- Steril:-
- Kapacitet:3,3 ml
- Beskrivelse:Extended fine tip
- Længde:153
- Seq:05
- Dråbevol.:20 µl