Transfer pipettes, fine tip
Ultra micro fine tip, 1,0 ml, 22 µl, 51 mm, -, Bulk
Katalog # 612-1738
Leverandør: Alpha Laboratories
Gå til:
Product Details & Documents
Non toxic, food grade LDPE.
- For careful control and small volume transfers
- Ideal for supernatant removal, slide preparation and gel loading
Documents
Specifikationer
- Pakket:Bulk
- Steril:-
- Kapacitet:1,0 ml
- Beskrivelse:Ultra micro fine tip
- Længde:51 mm
- Seq:02
- Dråbevol.:22 µl