Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for centrifuges

Accessories for centrifuges

Adapter for 30×15 ml tube, 17,5×74 mm
Katalog # 523-1482
Leverandør:   Hettich
Accessories for centrifuges
Katalog # 523-1482
Leverandør:  Hettich
Accessories for centrifuges
Accessories for centrifuges

Adapter for 30×15 ml tube, 17,5×74 mm

Price
1.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 30×15 ml tube, 17,5×74 mm
  • Seq:
    03
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.