Stirring rods
Omrørerstave, glas, med indsmeltede ender, Glas, 4 mm, 150 mm
Catalog # 441-9581
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
AR-Glas® with fused ends.
Documents
SCERL143000700915_TDS.pdf
Datasheet
Specifications
- Ø:4 mm
- Beskrivelse:Omrørerstave, glas, med indsmeltede ender
- Længde:150 mm
- Materiale:Glas
- Seq:01