Orders LinkContactUsLinkComponent
Scoops
Scoops
900 ml, 350 mm
Katalog # 231-0307
Leverandør:   SCHROEDER WILESCO
Scoops
Katalog # 231-0307
Leverandør:  SCHROEDER WILESCO
Scoops
Scoops
900 ml, 350 mm
Price
124,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifikationer
  • Kapacitet:
    900 ml
  • Længde:
    350 mm