Orders LinkContactUsLinkComponent
Multipurpose container, round, with snap-on lid
Multipurpose container, round, with snap-on lid
250 ml, 78,00 mm, 88,00 mm
Katalog # 216-3740
Leverandør:   hünersdorff GmbH
Multipurpose container, round, with snap-on lid
Katalog # 216-3740
Leverandør:  hünersdorff GmbH
Multipurpose container, round, with snap-on lid
Multipurpose container, round, with snap-on lid
250 ml, 78,00 mm, 88,00 mm
Price
32,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
HDPE, translucent.
  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable
  • Stackable
For storing and transporting solid and liquid materials, samples and general labware.
Documents
Specifikationer
  • Udv.Ø:
    78,00 mm
  • Kapacitet:
    250 ml
  • Højde:
    88,00 mm
  • Seq:
    01