Transport and Storage Boxes
42 l, Boks, opbevaring og transport, 550×350×220 mm
Katalog # 216-2121
Leverandør: ZARGES LEICHTBAU
Gå til:
Product Details & Documents
Aluminium alloy.
- Lid with stainless steel strap hinge, two straps and all-round seal
- Flip lock latches, secured with anti-tamper seals or padlock
- Flip handles on the front surfaces
Secure transportation, practical storage and protective packaging in one. The transported or stored item is securely packed and protected against external influences. Sturdy edge, lid and base profile. Stackable.
Specifikationer
- Kapacitet:42 l
- Beskrivelse:Boks, opbevaring og transport
- Seq:01
- B×D×H indv.:550×350×220 mm



