Orders LinkContactUsLinkComponent
Wire baskets
Wire baskets
180×180×180 mm
Katalog # 215-7418
Leverandør:   HUPFER METALLWERKE
Wire baskets
Katalog # 215-7418
Leverandør:  HUPFER METALLWERKE
Wire baskets
Wire baskets
180×180×180 mm
Price
1.200,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges
These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.
Specifikationer
  • Seq:
    06
  • B×D×H:
    180×180×180 mm