Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
48 (4×12), Stål, PA-belagt, 250×90×68 mm
Katalog # 212-6430
Leverandør:   HUPFER METALLWERKE
Racks for test tubes
Katalog # 212-6430
Leverandør:  HUPFER METALLWERKE
Racks for test tubes
Racks for test tubes
48 (4×12), Stål, PA-belagt, 250×90×68 mm
Price
708,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.
  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean
Trådracks med to etager, højde 21 mm. Samlet højde 68 mm. Hulstørrelse 18×18 mm.
Specifikationer
  • Ant. huller:
    48 (4×12)
  • Materiale:
    Stål, PA-belagt
  • Seq:
    15
  • B×D×H:
    250×90×68 mm