Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Racks for test tubes

Racks for test tubes

11 mm, 48 (4×12), 255×82×35 mm
Katalog # 212-6315
Racks for test tubes
Katalog # 212-6315
Racks for test tubes
Racks for test tubes

11 mm, 48 (4×12), 255×82×35 mm

Price
2.920,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Rack for test tubes, made from clear acrylic, constructed in two tiers.

  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
Specifikationer
  • Hul Ø:
    11 mm
  • Ant. huller:
    48 (4×12)
  • Seq:
    03
  • B×D×H:
    255×82×35 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.