Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for centrifuges
Siden oplever i øjeblikket tekniske problemer. Prøv venligst igen senere. Vi beklager ulejligheden.

Accessories for centrifuges

Adapter for 1×50 ml conical Falcon® tube (29×115 mm)
Katalog # 212-3611
   Lav fremstillingspåvirkning
Leverandør:   Hettich
Accessories for centrifuges
Katalog # 212-3611
Leverandør:  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter for 1×50 ml conical Falcon® tube (29×115 mm)

Price
1.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 1×50 ml conical Falcon® tube (29×115 mm)
  • Seq:
    16
  • Cat. No.:
    212-3611
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.