Orders LinkContactUsLinkComponent
Water purification systems, Super-Q® Plus
Water purification systems, Super-Q® Plus
OrangeX-Q cartridge, 22 inches, super-Q systems, ion-exchange, activated carbon
Katalog # 172-5058
Leverandør:   Merck
Water purification systems, Super-Q® Plus
Katalog # 172-5058
Leverandør:  Merck
Water purification systems, Super-Q® Plus
Water purification systems, Super-Q® Plus
OrangeX-Q cartridge, 22 inches, super-Q systems, ion-exchange, activated carbon
Price
7.650,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Super-Q® Plus System is a high-flow water system designed to provide the 'final polish' to water which has been pretreated by reverse osmosis, distillation or deionization. The systems consistently produce ultrapure water quality which exceeds Type 1 ASTM/CAP/CLSI specifications.
  • Modular design for the production of ultrapure water
  • Hourly automatic recirculation of water to maintain water quality between uses
  • Super-Q® water is produced 'on demand', so there is no need to store water and risk degrading its quality over time
Typical applications include: Tissue culture media preparation, reagent preparation, microelectronics wafer rinsing, microbiological buffer and media preparation.
Certifications: Exceeds Type 1 ASTM/CAP/CLSI specifications.
Specifikationer
  • Beskrivelse:
    OrangeX-Q cartridge, 22 inches, super-Q systems, ion-exchange, activated carbon
  • Seq:
    05