Laboratoriebørster
Bjørnehår, 12/4 mm
Katalog # 149-2153
Marketsource
Leverandør: VEREINIGTE PINSELFABRIKEN
Gå til:
Product Details & Documents
This brush range covers a wide variety of applications thanks to the number of styles on offer.
- Paintbrush made of pale ox hair
- Red marten brush — pure marten hair with black painted wooden handle
- Goat-hair brush — oval, on wooden handle
- Wide bear-hair brush, flat handle
Set 1: Contains 1 red marten brush in each size, sizes 1 to 5.
Specifikationer
- Børstemateriale:Bjørnehår
- Børstehoved Ø:12/4 mm
- Seq:11