Orders LinkContactUsLinkComponent
Laboratoriebørster
Laboratoriebørster
Size 6, Rødt mårhår, 3,8 mm
Katalog # 149-2126
Marketsource
Laboratoriebørster
Katalog # 149-2126
Marketsource
Laboratoriebørster
Laboratoriebørster
Size 6, Rødt mårhår, 3,8 mm
Price
67,51 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This brush range covers a wide variety of applications thanks to the number of styles on offer.
  • Paintbrush made of pale ox hair
  • Red marten brush — pure marten hair with black painted wooden handle
  • Goat-hair brush — oval, on wooden handle
  • Wide bear-hair brush, flat handle
Set 1: Contains 1 red marten brush in each size, sizes 1 to 5.
Specifikationer
  • Børstemateriale:
    Rødt mårhår
  • Børstehoved Ø:
    3,8 mm
  • Beskrivelse:
    Size 6
  • Seq:
    07