Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Latex gloves, Rotiprotect®

L, ufarvet, 240 mm, 0,20 mm, 100/box
  112-0042
Marketsource
 :   Roth Carl
Latex gloves, Rotiprotect®
  112-0042
 :  Roth Carl
Marketsource
Latex gloves, Rotiprotect®

L, ufarvet, 240 mm, 0,20 mm, 100/box

 
 
   
1 * 100 Items
 

These disposable gloves are suitable for protection against complex risks and for food processing applications.

  • Powder-free, smooth finish
  • Very smooth and pleasantly soft
  • Extremely stretchy
  • High tactile sensitivity
  • Thiuram-free

 : EN 374; Cat. III, AQL 1,5
Food contact approved

 
  • Handske størrelse:
    L
  • Pakket:
    100/box
  • Farve:
    ufarvet
  • Længde:
    240 mm
  • Seq:
    03
  • Tykkelse:
    0,20 mm
  • Cat. No.:
    112-0042
  • Ambidextrous/Hånd-specifik:
    Ambidextrous
  • Godkendt til fødevarekontakt:
    +
  • Med pudder/pudderfri:
    Pudderfri
  • Latex/latex-fri:
    Latex
  • Manchet:
    Rullet kant
  • EN norm:
    EN 374
  • Overflade:
    Smooth
  • Engangs/Genbrugelig:
    Engangs
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.