Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Latex gloves, Rotiprotect®

S, ufarvet, 240 mm, 0,20 mm, 100/box
Katalog # 112-0040
Marketsource
Leverandør:   Roth Carl
Latex gloves, Rotiprotect®
Katalog # 112-0040
Leverandør:  Roth Carl
Marketsource
Latex gloves, Rotiprotect®

S, ufarvet, 240 mm, 0,20 mm, 100/box

Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

These disposable gloves are suitable for protection against complex risks and for food processing applications.

  • Powder-free, smooth finish
  • Very smooth and pleasantly soft
  • Extremely stretchy
  • High tactile sensitivity
  • Thiuram-free

 : EN 374; Cat. III, AQL 1,5
Food contact approved

Specifikationer
  • Handske størrelse:
    S
  • Pakket:
    100/box
  • Farve:
    ufarvet
  • Længde:
    240 mm
  • Seq:
    01
  • Tykkelse:
    0,20 mm
  • Cat. No.:
    112-0040
  • Ambidextrous/Hånd-specifik:
    Ambidextrous
  • Godkendt til fødevarekontakt:
    +
  • Med pudder/pudderfri:
    Pudderfri
  • Latex/latex-fri:
    Latex
  • Manchet:
    Rullet kant
  • EN norm:
    EN 374
  • Overflade:
    Smooth
  • Engangs/Genbrugelig:
    Engangs