Orders LinkContactUsLinkComponent
Substrate buffers, Pierce™
Substrate buffers, Pierce™
Pierce™ Stable Peroxide Substrate Buffer, 10X, 100 mL
Katalog # PIER34062
Leverandør:   Thermo Fisher Scientific
Substrate buffers, Pierce™
Katalog # PIER34062
CAS nummer:  
Substrate buffers, Pierce™
Substrate buffers, Pierce™
Pierce™ Stable Peroxide Substrate Buffer, 10X, 100 mL
CAS nummer:  
Price
778,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
These stable peroxide and diethanolamine substrate buffers produce optimised, reliable and convenient stock solutions for HRP and alkaline phosphatase assays detection respectively.
  • Ready to use
  • Minimises assay-to-assay variability
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    Pierce™ Stable Peroxide Substrate Buffer, 10X
Specifikationer