Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pierce™ Protein A

Pierce™ Protein A

Protein A, biotinylated, 1 mg
Katalog # PIER29989
Leverandør:   Thermo Fisher Scientific
Pierce™ Protein A
Katalog # PIER29989
CAS nummer:  
Pierce™ Protein A
Pierce™ Protein A

Protein A, biotinylated, 1 mg

CAS nummer:  
Price
1.710,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four Fc-binding domains per protein
  • Efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies
Specifikationer

Specifikationer

  • Pakn:
    1 mg
  • Beskrivelse:
    Protein A, biotinylated

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.