Orders LinkContactUsLinkComponent
Drying flasks
Drying flasks
2000 ml, 29/32 NS
Katalog # 531-1983
Leverandør:   BUCHI
Drying flasks
Katalog # 531-1983
Leverandør:  BUCHI
Drying flasks
Drying flasks
2000 ml, 29/32 NS
Price
1.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Made of Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rotary evaporators Rotavapor R-210/215. Pear shape with NS 29/32 neck. Glassware A, V, C.
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    2000 ml