Orders LinkContactUsLinkComponent
Dækglas, Corning® BioCoat™
Dækglas, Corning® BioCoat™
Dyrkningsskål, Poly-D-lysin-belagt, 12 mm, Glas
Katalog # 734-1006
   Lav fremstillingspåvirkning
Leverandør:   Corning
Dækglas, Corning® BioCoat™
Katalog # 734-1006
Leverandør:  Corning
   Lav fremstillingspåvirkning
Dækglas, Corning® BioCoat™
Dækglas, Corning® BioCoat™
Dyrkningsskål, Poly-D-lysin-belagt, 12 mm, Glas
Price
2.720,00 kr
Log ind for tilgængelighed
1 * 80 Items
Product Details & Documents
These cover glasses have a uniform coating of either poly-L-lysine, poly-D-lysine and laminin, human fibronectin or rat tail collagen.
  • Optically clear
  • Low background fluorescence
  • Non-neurotoxic
Designed for applications where optically clear, thin glass bottoms are needed such as immunofluorescence microscopy, fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy and phase contrast microscopy.
Documents
Specifikationer
  • Ø:
    12 mm
  • Beskrivelse:
    Dyrkningsskål, Poly-D-lysin-belagt
  • Materiale:
    Glas
  • Cat. No.:
    734-1006
  • Miljøcertificering:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Lav fremstillingspåvirkning