Orders LinkContactUsLinkComponent
Dækglas, Corning® BioCoat™
Dækglas, Corning® BioCoat™
Dyrkningsskål, Poly-L-lysin-belagt, 12 mm, Glas
Catalog # 734-1005
   Lav fremstillingspåvirkning
Supplier:   Corning
Dækglas, Corning® BioCoat™
Catalog # 734-1005
Supplier:  Corning
   Lav fremstillingspåvirkning
Dækglas, Corning® BioCoat™
Dækglas, Corning® BioCoat™
Dyrkningsskål, Poly-L-lysin-belagt, 12 mm, Glas
Price
2.820,00 kr
Log in for availability
1 * 80 Items
Product Details & Documents
These cover glasses have a uniform coating of either poly-L-lysine, poly-D-lysine and laminin, human fibronectin or rat tail collagen.
  • Optically clear
  • Low background fluorescence
  • Non-neurotoxic
Designed for applications where optically clear, thin glass bottoms are needed such as immunofluorescence microscopy, fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy and phase contrast microscopy.
Documents
Specifications
  • Ø:
    12 mm
  • Beskrivelse:
    Dyrkningsskål, Poly-L-lysin-belagt
  • Materiale:
    Glas
  • Cat. No.:
    734-1005
  • Miljøcertificering:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Lav fremstillingspåvirkning